Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027008.2 | NP_081284.2 | 1, 2 | 311 | |
GenBank mRNA | AK011637.1 | - | 1, 2 | 46 | |
AK052960.1 | - | 1, 2 | 313 | ||
AK129077.1 | - | 1, 2 | 271 | ||
AK143644.1 | - | 1, 2 | 279 | ||
AK187166.1 | - | 1, 2 | 28 | ||
BC021777.1 | - | 1, 2 | 263 | 4215964 |