Gene Transcripts

Gene Symbol
Cypt3
Entrez Gene ID
69361
Gene Name
cysteine-rich perinuclear theca 3
Gene Aliases
1700006H02Rik, Ckt1r1
Chromosome Location
Chr. X: 153558574 - 153559436 on Build GRCm38
UniGene ID
Mm.46095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173367.2 NP_775543.2 2 345
GenBank mRNA AF463500.1 - 2 325
AK005674.1 - 2 358
AM040452.1 - 2 340
BC048527.1 - 2 345 6772990

View Full Product Details