Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027117.3 | NP_081393.2 | 6 | 895 | |
GenBank mRNA | AK009481.1 | - | 6 | 810 | |
AK051418.1 | - | 6 | 822 | ||
AK164223.1 | - | 4 | 271 | ||
AK178282.1 | - | 4 | 281 | ||
AY513272.1 | - | 6 | 846 | ||
BC005581.1 | - | 6 | 819 | 3968615 |