Gene Transcripts

Gene Symbol
Chp2
Entrez Gene ID
70261
Gene Name
calcineurin-like EF hand protein 2
Gene Aliases
2010110P09Rik, Cbhp2
Chromosome Location
Chr. 7: 122208621 - 122222539 on Build GRCm38
UniGene ID
Mm.96997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027363.1 NP_081639.1 6 695
GenBank mRNA AK008392.1 - 6 529
BC119202.1 - 6 695 8733951
BC119204.1 - 6 695 8733953

View Full Product Details