Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
70314
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
2610011A08Rik, AI462746, Fra
Chromosome Location
Chr. 7: 126428416 - 126445907 on Build GRCm38
UniGene ID
Mm.35467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030566.2 NP_085043.2 11 1776
GenBank mRNA AK089938.1 - 11 1481
AK144836.1 - 11 1462
AK157832.1 - 11 1437
BC004088.1 - 10 1522 3593722
BC015287.1 - 11 1784 4240442

View Full Product Details