Gene Transcripts

Gene Symbol
Inf2
Entrez Gene ID
70435
Gene Name
inverted formin, FH2 and WH2 domain containing
Gene Aliases
2610204M08Rik, AA589465, AW125550, EG629699
Chromosome Location
Chr. 12: 112588746 - 112615557 on Build GRCm38
UniGene ID
Mm.250193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198411.2 NP_940803.2 6 917
GenBank mRNA CD347830.1 - 5 550
DQ834374.1 - 6 919

View Full Product Details