Gene Transcripts

Gene Symbol
Rasl12
Entrez Gene ID
70784
Gene Name
RAS-like, family 12
Gene Aliases
4631404I11Rik
Chromosome Location
Chr. 9: 65398485 - 65415060 on Build GRCm38
UniGene ID
Mm.35827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033158.2 NP_001028330.2 5 1804
GenBank mRNA AK014511.1 - 5 1767
AK171623.1 - 5 1760

View Full Product Details