Gene Transcripts

Gene Symbol
Arrdc2
Entrez Gene ID
70807
Gene Name
arrestin domain containing 2
Gene Aliases
4632416I05Rik, Ilad1
Chromosome Location
Chr. 8: 70835129 - 70839720 on Build GRCm38
UniGene ID
Mm.24215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027560.1 NP_081836.1 7 1416
GenBank mRNA AK014582.1 - 7 1416
AK046754.1 - 7 1126
AK135740.1 - 7 1232
AK137175.1 - 7 1233
AK144323.1 - 8 1150
BC034910.1 - 1 295
BC036384.1 - 7 1214 1067080
BC043313.1 - 2 936
BC043469.1 - 2 936
BC054826.1 - 2 942
BC094060.1 - 6 1296
DQ861995.1 - 7 1080

View Full Product Details