Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162979.1 | NP_001156451.1 | 12 | 1785 | |
GenBank mRNA | AK014876.1 | - | 12 | 1768 | |
AK029544.1 | - | 12 | 1784 | ||
BC127049.1 | - | 12 | 1772 | ||
BC152307.1 | - | 12 | 1774 | 40062409 | |
BC152308.1 | - | 12 | 1774 | 40062416 |