Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027764.2 | NP_082040.2 | 9 | 1527 | |
GenBank mRNA | AK033501.1 | - | 9 | 1461 | |
AK088299.1 | - | 9 | 1516 | ||
AK089923.1 | - | 10 | 1528 | ||
AK187668.1 | - | 2 | 110 | ||
BC040764.1 | - | 2 | 222 | ||
BC067005.1 | - | 10 | 1581 | 5721370 |