Gene Transcripts

Gene Symbol
Dhrs2
Entrez Gene ID
71412
Gene Name
dehydrogenase/reductase member 2
Gene Aliases
5430405K24Rik, Hep27
Chromosome Location
Chr. 14: 55124154 - 55241440 on Build GRCm38
UniGene ID
Mm.85484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027790.2 NP_082066.2 6 646
GenBank mRNA AK017274.1 - 6 648
BC117731.2 - 5 539 40089799

View Full Product Details