Gene Transcripts

Gene Symbol
Mettl13
Entrez Gene ID
71449
Gene Name
methyltransferase like 13
Gene Aliases
5630401D24Rik
Chromosome Location
Chr. 1: 162533671 - 162548345 on Build GRCm38
UniGene ID
Mm.270569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144877.1 NP_659126.1 8 2137
GenBank mRNA AK084750.1 - 8 1832
AK146751.1 - 2335
AK168959.1 - 8 2344
AK169319.1 - 2336
AK173044.1 - 2290
BC014872.1 - 8 2137 3602469

View Full Product Details