Gene Transcripts

Gene Symbol
Bbs7
Entrez Gene ID
71492
Gene Name
Bardet-Biedl syndrome 7 (human)
Gene Aliases
8430406N16Rik
Chromosome Location
Chr. 3: 36573140 - 36613489 on Build GRCm38
UniGene ID
Mm.286187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027810.3 NP_082086.2 18 2011
GenBank mRNA AF521645.1 - 18 2010
AK132661.1 - 18 1939
BC031505.1 - 18 1961 3494587

View Full Product Details