Gene Transcripts

Gene Symbol
Arl14
Entrez Gene ID
71619
Gene Name
ADP-ribosylation factor-like 14
Gene Aliases
9130014L17Rik, Arf7
Chromosome Location
Chr. 3: 69222419 - 69223618 on Build GRCm38
UniGene ID
Mm.98122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027843.1 NP_082119.1 1 198
GenBank mRNA AK018621.1 - 1 199
BC104368.1 - 1 159 40049874
BC104369.1 - 1 159 40049875

View Full Product Details