Gene Transcripts

Gene Symbol
Rarres2
Entrez Gene ID
71660
Gene Name
retinoic acid receptor responder (tazarotene induced) 2
Gene Aliases
0610007L05Rik, AI303516
Chromosome Location
Chr. 6: 48569696 - 48572844 on Build GRCm38
UniGene ID
Mm.28231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027852.2 NP_082128.1 3 371
GenBank mRNA AK002298.1 - 3 372
BC015275.1 - 2 551
BC038914.1 - 3 391 5100583

View Full Product Details