Gene Transcripts
- Gene Symbol
- Fuca1
- Entrez Gene ID
- 71665
- Gene Name
- fucosidase, alpha-L- 1, tissue
- Gene Aliases
- 0610006A03Rik, 9530055J05Rik, Afuc, Fuca
- Chromosome Location
- Chr. 4: 135920726 - 135940300 on Build GRCm38
- UniGene ID
- Mm.439940
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024243.4 |
NP_077205.3
|
4, 5 |
797 |
|
GenBank mRNA |
AK002230.1 |
- |
4, 5 |
765 |
|
AK053323.1 |
- |
4, 5 |
742 |
|
AK089958.1 |
- |
4, 5 |
767 |
|
AK150526.1 |
- |
4, 5 |
732 |
|
AK150890.1 |
- |
4, 5 |
787 |
|
AK151258.1 |
- |
4, 5 |
765 |
|
AK151361.1 |
- |
4, 5 |
786 |
|
AK151879.1 |
- |
4, 5 |
786 |
|
AK151908.1 |
- |
4, 5 |
740 |
|
AK152070.1 |
- |
4, 5 |
740 |
|
AK165604.1 |
- |
4, 5 |
767 |
|
AK170503.1 |
- |
4, 5 |
777 |
|
BC003235.1 |
- |
4, 5 |
770 |
3497631 |
View Full Product Details