Gene Transcripts

Gene Symbol
Pnmal1
Entrez Gene ID
71691
Gene Name
PNMA-like 1
Gene Aliases
0710005I19Rik
Chromosome Location
Chr. 7: 16949837 - 16964607 on Build GRCm38
UniGene ID
Mm.275673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007569.1 NP_001007570.1 2 919
GenBank mRNA BC048228.1 - 2 919

View Full Product Details