Gene Transcripts

Gene Symbol
Osbpl3
Entrez Gene ID
71720
Gene Name
oxysterol binding protein-like 3
Gene Aliases
1200014M06Rik, 6720421I08Rik, A530055M08, ORP3, OSBP3
Chromosome Location
Chr. 6: 50293326 - 50456355 on Build GRCm38
UniGene ID
Mm.44153
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163645.1 NP_001157117.1 13 1848
NM_027881.3 NP_082157.2 13 1566
GenBank mRNA AK004768.1 - 13 1568
AK037114.1 - 13 1535
AK173006.1 - 14 4780
BC044866.1 - 12 1493 5003631
BC086485.1 - 13 1760 5344992

View Full Product Details