Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027889.1 | NP_082165.1 | 4 | 596 | |
GenBank mRNA | AK004695.1 | - | 4 | 571 | |
AK032484.1 | - | 4 | 561 | ||
AK037658.1 | - | 4 | 570 | ||
AK047228.1 | - | 4 | 404 | ||
AK090030.1 | - | 4 | 577 | ||
AK153963.1 | - | 4 | 569 | ||
AK162897.1 | - | 4 | 570 | ||
AK182905.1 | - | 1 | 35 | ||
AK196649.1 | - | 3 | 341 | ||
BC029004.1 | - | 4 | 596 | 5360111 |