Gene Transcripts

Gene Symbol
Tmem50a
Entrez Gene ID
71817
Gene Name
transmembrane protein 50A
Gene Aliases
3200001F09Rik, CAM, Smp1
Chromosome Location
Chr. 4: 134897849 - 134915030 on Build GRCm38
UniGene ID
Mm.88349
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027935.2 NP_082211.1 3, 4 254
GenBank mRNA AK014282.1 - 3, 4 324
AK147069.1 - 3, 4 290
AK152630.1 - 3, 4 338
AK152654.1 - 3, 4 309
AK157214.1 - 3, 4 327
AK164989.1 - 3, 4 320
AK165636.1 - 3, 4 303
BC094256.1 - 3, 4 254 6820666

View Full Product Details