Gene Transcripts
- Gene Symbol
- MTAP
- Entrez Gene ID
- 4507
- Gene Name
- methylthioadenosine phosphorylase
- Gene Aliases
- BDMF, DMSFH, DMSMFH, HEL-249, LGMBF, MSAP, c86fus
- Chromosome Location
- Chr. 9: 21802636 - 21865971 on Build GRCh38
- UniGene ID
- Hs.193268
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002451.3 |
NP_002442.2
|
5, 6 |
564 |
|
GenBank mRNA |
AK300592.1 |
- |
5, 6 |
727 |
|
AK309365.1 |
- |
5, 6 |
551 |
|
AY712791.1 |
- |
5, 6 |
572 |
|
BC012316.1 |
- |
5, 6 |
538 |
3546198 |
BC026106.1 |
- |
5, 6 |
546 |
4820938 |
CR541670.1 |
- |
5, 6 |
451 |
|
CR541710.1 |
- |
5, 6 |
451 |
|
FJ224338.1 |
- |
5, 6 |
451 |
|
HE654772.1 |
- |
5, 6 |
560 |
|
HE654773.1 |
- |
5, 6 |
560 |
|
HE654774.1 |
- |
5, 6 |
560 |
|
HE654775.1 |
- |
5, 6 |
560 |
|
HE654776.1 |
- |
5, 6 |
560 |
|
HE654777.1 |
- |
5, 6 |
560 |
|
L40432.1 |
- |
5, 6 |
561 |
|
U22233.1 |
- |
5, 6 |
572 |
|
View Full Product Details