Gene Transcripts

Gene Symbol
Coq2
Entrez Gene ID
71883
Gene Name
coenzyme Q2 homolog, prenyltransferase (yeast)
Gene Aliases
2310002F18Rik, PHB:PPT
Chromosome Location
Chr. 5: 100654723 - 100674285 on Build GRCm38
UniGene ID
Mm.260661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027978.2 NP_082254.2 2 346
GenBank mRNA AK009092.1 - 2 348
BC080773.1 - 2 341 30139077

View Full Product Details