Gene Transcripts

Gene Symbol
Zcchc24
Entrez Gene ID
71918
Gene Name
zinc finger, CCHC domain containing 24
Gene Aliases
2310047A01Rik
Chromosome Location
Chr. 14: 25711640 - 25768856 on Build GRCm38
UniGene ID
Mm.41904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101433.1 NP_001094903.1 2 262
GenBank mRNA BC147263.1 - 2 352
BC147264.1 - 2 352 9056314
CN526234.1 - 2 42

View Full Product Details