Gene Transcripts
- Gene Symbol
- Seh1l
- Entrez Gene ID
- 72124
- Gene Name
- SEH1-like (S. cerevisiae
- Gene Aliases
- 2610007A16Rik, AW540070, SEC13L, SEH1A, SEH1B, Seh1
- Chromosome Location
- Chr. 18: 67774798 - 67795487 on Build GRCm38
- UniGene ID
- Mm.307315
- Species
- Mus musculus
- Gene Symbol
- 4930549G23Rik
- Entrez Gene ID
- 75286
- Gene Name
- RIKEN cDNA 4930549G23 gene
- Gene Aliases
- Chromosome Location
- Chr. 18: 67776033 - 67799943 on Build GRCm38
- UniGene ID
- Mm.307315
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CN666923.1 |
- |
1 |
44 |
|
View Full Product Details