Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028128.1 | NP_082404.1 | 6, 7 | 629 | |
GenBank mRNA | AK011489.1 | - | 6, 7 | 640 | |
AK150036.1 | - | 642 | |||
AK170211.1 | - | 6, 7 | 647 | ||
AK171372.1 | - | 629 | |||
BC023674.1 | - | 5, 6 | 567 | ||
BC089001.1 | - | 6, 7 | 629 | 6415420 |