Gene Transcripts
- Gene Symbol
- Cyth4
- Entrez Gene ID
- 72318
- Gene Name
- cytohesin 4
- Gene Aliases
- 2510004M07Rik, 5830469K17Rik, AI467541, Pscd4, mFLJ00017
- Chromosome Location
- Chr. 15: 78597047 - 78622019 on Build GRCm38
- UniGene ID
- Mm.32911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028195.3 |
NP_082471.2
|
8, 9 |
853 |
|
GenBank mRNA |
AF079972.1 |
- |
8, 9 |
784 |
|
AK030956.1 |
- |
8, 9 |
1738 |
|
AK152306.1 |
- |
8, 9 |
759 |
|
AK152909.1 |
- |
8, 9 |
759 |
|
AK157026.1 |
- |
8, 9 |
759 |
|
AK220198.1 |
- |
8, 9 |
782 |
|
BC018505.1 |
- |
7, 8 |
677 |
|
BC039640.1 |
- |
1, 2 |
272 |
|
BC139054.1 |
- |
8, 9 |
826 |
8862076 |
BC139056.1 |
- |
8, 9 |
826 |
8862078 |
BC145528.1 |
- |
8, 9 |
853 |
|
View Full Product Details