Gene Transcripts

Gene Symbol
Psca
Entrez Gene ID
72373
Gene Name
prostate stem cell antigen
Gene Aliases
2210408B04Rik
Chromosome Location
Chr. 15: 74714839 - 74717065 on Build GRCm38
UniGene ID
Mm.46395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028216.2 NP_082492.1 2 122
GenBank mRNA AF319173.1 - 2 110
AK008851.1 - 2 122
BC110462.2 - 2 122 40039785
EH109693.1 - 2 119

View Full Product Details