Gene Transcripts

Gene Symbol
Ier5l
Entrez Gene ID
72500
Gene Name
immediate early response 5-like
Gene Aliases
2610524G09Rik
Chromosome Location
Chr. 2: 30472631 - 30474199 on Build GRCm38
UniGene ID
Mm.295464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030244.3 NP_084520.3 1 1246
GenBank mRNA AK012145.1 - 1 1259
AK034055.1 - 1269
BC003464.1 - 1 1245 2648900
BC010234.1 - 1 1246 3499467
BC040084.1 - 1 1246 3499467

View Full Product Details