Gene Transcripts

Gene Symbol
Tmem55a
Entrez Gene ID
72519
Gene Name
transmembrane protein 55A
Gene Aliases
2610319K07Rik, AI315591, AV001360
Chromosome Location
Chr. 4: 14864219 - 14915260 on Build GRCm38
UniGene ID
Mm.279158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028264.4 NP_082540.1 6 711
GenBank mRNA AK012054.1 - 6 671
AK033845.1 - 6 674
AK049213.1 - 6 712
AX775959.1 - 6 640
BC021435.1 - 6 656 5029766

View Full Product Details