Gene Transcripts

Gene Symbol
Nfatc4
Entrez Gene ID
73181
Gene Name
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4
Gene Aliases
3110041H08Rik, AW107667, AW546455, Nfat3
Chromosome Location
Chr. 14: 55824795 - 55833943 on Build GRCm38
UniGene ID
Mm.27908
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023699.3 NP_076188.3 9, 10 2884
GenBank mRNA AK159078.1 - 9, 10 2730
BC028928.1 - 9, 10 2671 3670623
EU887656.1 - 9, 10 2642
EU887658.1 - 4, 5 1097
EU887660.1 - 3, 4 2153

View Full Product Details