Gene Transcripts

Gene Symbol
Prss52
Entrez Gene ID
73382
Gene Name
protease, serine 52
Gene Aliases
1700049K14Rik, Tesp3
Chromosome Location
Chr. 14: 64104323 - 64113751 on Build GRCm38
UniGene ID
Mm.481055
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028525.2 NP_082801.2 4 672
GenBank mRNA AB009660.1 - 4 709
AK006746.1 - 4 671
BC049714.1 - 4 672 6741678

View Full Product Details