Gene Transcripts

Gene Symbol
Gucy2g
Entrez Gene ID
73707
Gene Name
guanylate cyclase 2g
Gene Aliases
2410077I05Rik, GC-G
Chromosome Location
Chr. 19: 55198098 - 55242429 on Build GRCm38
UniGene ID
Mm.489987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081076.2 NP_001074545.1 4 952
GenBank mRNA AY395631.1 - 4 952

View Full Product Details