Gene Transcripts

Gene Symbol
Lce1c
Entrez Gene ID
73719
Gene Name
late cornified envelope 1C
Gene Aliases
1110014K05Rik
Chromosome Location
Chr. 3: 92679247 - 92680918 on Build GRCm38
UniGene ID
Mm.292458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028622.2 NP_082898.1 2 637
GenBank mRNA AK003705.1 - 2 638

View Full Product Details