Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134471.4 | NP_608301.3 | 2 | 270 | |
GenBank mRNA | AK015046.1 | - | 2 | 240 | |
AK088047.1 | - | 2 | 211 | ||
AK150222.1 | - | 2 | 201 | ||
BC006841.1 | - | 2 | 230 | 3598157 | |
CF163926.1 | - | 2 | 270 |