Gene Transcripts

Gene Symbol
Atl1
Entrez Gene ID
73991
Gene Name
atlastin GTPase 1
Gene Aliases
4930435M24Rik, Adfsp, Fsp1, Spg3, Spg3a
Chromosome Location
Chr. 12: 69893105 - 69964085 on Build GRCm38
UniGene ID
Mm.474462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178628.5 NP_848743.1 8 1058
GenBank mRNA AK015327.1 - 1 70
AK046919.1 - 7 788
AK050339.1 - 8 895
AK140185.1 - 7 802
AK147311.1 - 7 4096
BC050784.1 - 7 787 6595181
BC057068.1 - 3 619 5689726

View Full Product Details