Gene Transcripts
- Gene Symbol
- Syvn1
- Entrez Gene ID
- 74126
- Gene Name
- synovial apoptosis inhibitor 1, synoviolin
- Gene Aliases
- 1200010C09Rik, AW211966, C85322, D530017H19Rik, Hrd1
- Chromosome Location
- Chr. 19: 6045030 - 6053718 on Build GRCm38
- UniGene ID
- Mm.149870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164709.1 |
NP_001158181.1
|
5, 6 |
631 |
|
NM_028769.5 |
NP_083045.4
|
5, 6 |
998 |
|
GenBank mRNA |
AK004688.2 |
- |
5, 6 |
485 |
|
AK122558.1 |
- |
4, 5 |
600 |
|
AK143720.1 |
- |
5, 6 |
510 |
|
AK172097.1 |
- |
4, 5 |
597 |
|
BC042199.1 |
- |
5, 6 |
493 |
4953165 |
BC046829.1 |
- |
5, 6 |
631 |
6811574 |
BC057917.1 |
- |
5, 6 |
912 |
5136398 |
BC080722.1 |
- |
5, 6 |
516 |
5323695 |
View Full Product Details