Gene Transcripts

Gene Symbol
Smu1
Entrez Gene ID
74255
Gene Name
smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans)
Gene Aliases
2600001O03Rik, 2610203K23Rik, AB044414, AI845086, AW556129, Bwd
Chromosome Location
Chr. 4: 40735649 - 40757885 on Build GRCm38
UniGene ID
Mm.289929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021535.4 NP_067510.3 9, 10 1208
GenBank mRNA AB044414.1 - 9, 10 1137
AK003493.1 - 9, 10 1195
AK019174.1 - 9, 10 1167
AK145982.1 - 9, 10 1207
AK159635.1 - 9, 10 1195
AK168813.1 - 9, 10 1202
AK212788.1 - 13
BC020034.1 - 9, 10 1150
BC057446.1 - 9, 10 1169 3155760

View Full Product Details