Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004539.3 | NP_004530.1 | 13 | 1667 | |
XM_005266700.1 | XP_005266757.1 | 13 | 1445 | ||
GenBank mRNA | AJ000334.1 | - | 13 | 1512 | |
AK129897.1 | - | 13 | 1469 | ||
AK222835.1 | - | 13 | 1514 | ||
AK297777.1 | - | 11 | 1098 | ||
BC001687.1 | - | 13 | 1516 | 2987840 | |
D84273.1 | - | 13 | 1471 |