Gene Transcripts

Gene Symbol
Fam53a
Entrez Gene ID
74504
Gene Name
family with sequence similarity 53, member A
Gene Aliases
2410018C17Rik, 5430419M09Rik, Dntnp
Chromosome Location
Chr. 5: 33600351 - 33629647 on Build GRCm38
UniGene ID
Mm.294695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178390.3 NP_848477.2 5 1462
GenBank mRNA AK010537.1 - 5 1398
AK017321.1 - 4 1807
AK155077.1 - 6 1575
AK163326.1 - 5 1434
AK220276.1 - 4 1098
BC016596.1 - 3 887 3257498
BC028656.1 - 5 1182 1244982

View Full Product Details