Gene Transcripts

Gene Symbol
Ttc17
Entrez Gene ID
74569
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
9130020K17Rik, D2Bwg1005e
Chromosome Location
Chr. 2: 94300766 - 94406689 on Build GRCm38
UniGene ID
Mm.248504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183106.2 NP_898929.2 11 1387
GenBank mRNA AK197793.1 - 2 127
BY358550.1 - 3 206

View Full Product Details