Gene Transcripts

Gene Symbol
Wdr35
Entrez Gene ID
74682
Gene Name
WD repeat domain 35
Gene Aliases
4930459M12Rik, 4931430C06, mKIAA1336
Chromosome Location
Chr. 12: 8973992 - 9028848 on Build GRCm38
UniGene ID
Mm.87389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159527.1 NP_001152999.1 27 3561
NM_172470.3 NP_766058.3 28 3594
GenBank mRNA AK029896.1 - 27 3516
AK083971.1 - 27 3506
BC056925.1 - 28 3703 5704780

View Full Product Details