Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000433.3 | NP_000424.2 | 2 | 471 | |
NM_001127651.2 | NP_001121123.1 | 3 | 308 | ||
NM_001190789.1 | NP_001177718.1 | 2 | 471 | ||
NM_001190794.1 | NP_001177723.1 | 2 | 471 | ||
XM_005245207.1 | XP_005245264.1 | 2 | 400 | ||
XM_011509580.1 | XP_011507882.1 | 3 | 309 | ||
XM_011509581.1 | XP_011507883.1 | 3 | 296 | ||
GenBank mRNA | AB209647.1 | - | 2 | 351 | |
AF527950.1 | - | 2 | 218 | ||
AK296672.1 | - | 2 | 327 | ||
AK298713.1 | - | 2 | 380 | ||
AK312666.1 | - | 2 | 381 | ||
BC001606.1 | - | 2 | 448 | 3542841 | |
BP193903.1 | - | 3 | 155 | ||
BT007439.1 | - | 2 | 196 | ||
DA420141.1 | - | 3 | 299 | ||
DA817767.1 | - | 3 | 309 | ||
DC358267.1 | - | 2 | 471 | ||
KU178216.1 | - | 2 | 196 | ||
M32011.1 | - | 3 | 263 |