Gene Transcripts
- Gene Symbol
- Iqca
- Entrez Gene ID
- 74918
- Gene Name
- IQ motif containing with AAA domain
- Gene Aliases
- 4930465P12Rik, 4930585L22Rik, BB288505, Iqca1
- Chromosome Location
- Chr. 1: 90042132 - 90153401 on Build GRCm38
- UniGene ID
- Mm.317517
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029122.2 |
NP_083398.2
|
16, 17 |
2182 |
|
GenBank mRNA |
AK139726.1 |
- |
9, 10 |
1037 |
|
View Full Product Details