Gene Transcripts

Gene Symbol
NDUFB6
Entrez Gene ID
4712
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
B17, CI
Chromosome Location
Chr. 9: 32553526 - 32573184 on Build GRCh38
UniGene ID
Hs.493668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002493.4 NP_002484.1 2 371
NM_182739.2 NP_877416.1 2 371
GenBank mRNA AF035840.1 - 2 315
AF067167.1 - 2 344
AK289702.1 - 2 325
BC009801.1 - 2 350 4334414
BC016708.1 - 2 385
BC029247.1 - 2 316 5171627
BI669356.1 - 2 350
BP208846.1 - 2 369
CR456920.1 - 2 247

View Full Product Details