Gene Transcripts

Gene Symbol
Wnk2
Entrez Gene ID
75607
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
1810073P09Rik, AW122246, ESTM15, X83337, mKIAA1760
Chromosome Location
Chr. 13: 49036302 - 49148574 on Build GRCm38
UniGene ID
Mm.342723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290311.1 NP_001277240.1 3, 4 1416
NM_001290313.1 NP_001277242.1 3, 4 1416
NM_029361.4 NP_083637.2 3, 4 1416
GenBank mRNA AK147550.1 - 3, 4 1103

View Full Product Details