Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029364.3 | NP_083640.1 | 3 | 406 | |
GenBank mRNA | AK030773.1 | - | 3 | 405 | |
AK049162.1 | - | 3 | 407 | ||
AK054046.1 | - | 3 | 406 | ||
AK083597.1 | - | 3 | 384 | ||
AK149863.1 | - | 3 | 391 | ||
AK151037.1 | - | 1 | 28 | ||
AK159562.1 | - | 3 | 373 | ||
AK165180.1 | - | 3 | 386 | ||
AK165917.1 | - | 3 | 356 | ||
AK169485.1 | - | 3 | 388 | ||
AK170791.1 | - | 3 | 402 | ||
BC055328.1 | - | 3 | 400 | 6400020 |