Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164640.1 | NP_001158112.1 | 5 | 1256 | |
NM_029419.2 | NP_083695.2 | 6 | 1356 | ||
GenBank mRNA | AK018646.1 | - | 6 | 1358 | |
AK085956.1 | - | 5 | 1233 | ||
BC139137.1 | - | 5 | 1135 | 8862159 | |
BC139138.1 | - | 5 | 1135 | 8862160 |