Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289506.1 | NP_001276435.1 | 3 | 1213 | |
NM_029422.4 | NP_083698.1 | 3 | 1216 | ||
GenBank mRNA | AB109560.1 | - | 3 | 1214 | |
AK154384.1 | - | 3 | 1215 | ||
AY517483.1 | - | 3 | 1215 | ||
AY517484.1 | - | 4 | 966 | ||
BC139146.1 | - | 3 | 1202 | 8862168 | |
BC145558.1 | - | 3 | 1199 | 9054122 |