Gene Transcripts
- Gene Symbol
- Mgea5
- Entrez Gene ID
- 76055
- Gene Name
- meningioma expressed antigen 5 (hyaluronidase)
- Gene Aliases
- 2810009A20Rik, 4833427O07Rik, 5830447M11Rik, AA408215, Hy5, Ncoat, OGA, mKIAA0679
- Chromosome Location
- Chr. 19: 45750255 - 45783832 on Build GRCm38
- UniGene ID
- Mm.122725
- Species
- Mus musculus
- Gene Symbol
- D19Ertd409e
- Entrez Gene ID
- 52016
- Gene Name
- DNA segment, Chr 19, ERATO Doi 409, expressed
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.122725
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK163010.1 |
- |
1 |
1766 |
|
BG080263.2 |
- |
1 |
243 |
|
View Full Product Details