Gene Transcripts

Gene Symbol
Mcur1
Entrez Gene ID
76137
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
6230416A05Rik, AU015498, AV136929, AW554392, C88263, Ccdc90a
Chromosome Location
Chr. 13: 43538406 - 43560259 on Build GRCm38
UniGene ID
Mm.426290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081059.3 NP_001074528.1 8 1012
GenBank mRNA AK018089.1 - 8 983
AK186531.1 - 3 203
AK210359.1 - 3 203
BC113195.1 - 9 750

View Full Product Details